Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004578.3 | NP_004569.2 | 3, 4 | 513 | ||
NR_073545.1 | 3, 4 | 513 | ||||
GenBank mRNA | AF498934.1 | - | 3, 4 | 235 | ||
AK223009.1 | - | 3, 4 | 278 | |||
AK313807.1 | - | 3, 4 | 486 | |||
AY585832.1 | - | 3, 4 | 442 | |||
BC002438.1 | - | 3, 4 | 306 | 3346455 | ||
BC004309.1 | - | 3, 4 | 443 | 3627110 | ||
BI546021.1 | - | 3, 4 | 517 | |||
BI828974.1 | - | 3, 4 | 397 | |||
KC407675.1 | - | 3, 4 | 235 | |||
M28211.1 | - | 3, 4 | 290 | |||
X13902.1 | - | 3, 4 | 346 |