Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289807.1 | NP_001276736.1 | 3 | 542 | ||
NM_001289808.1 | NP_001276737.1 | 2 | 571 | |||
NM_001885.2 | NP_001876.1 | 3 | 576 | |||
XM_011542608.1 | XP_011540910.1 | 3 | 557 | |||
XM_011542609.2 | XP_011540911.1 | 2 | 798 | |||
GenBank mRNA | AF007162.1 | - | 2 | 298 | ||
AK295498.1 | - | 2 | 361 | |||
AK314029.1 | - | 2 | 314 | |||
BC007008.1 | - | 2 | 315 | 3933748 | ||
BI759672.1 | - | 3 | 486 | |||
BM721803.1 | - | 3 | 537 | |||
BT006770.1 | - | 2 | 269 | |||
FJ224314.1 | - | 2 | 294 | |||
FJ876064.1 | - | 2 | 269 | |||
KM220603.1 | - | 2 | 283 | |||
KM220604.1 | - | 2 | 283 | |||
KM220605.1 | - | 2 | 283 | |||
KM220606.1 | - | 2 | 293 | |||
KM220607.1 | - | 2 | 248 | |||
S45630.1 | - | 2 | 294 |