Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALPK1 SIRNA Assays ›
Gene Symbol:
ALPK1
Gene Name:
alpha kinase 1
Gene Aliases:
8430410J10Rik, LAK
Chromosome Location:
Chr.4: 112286420 - 112442618 on Build GRCh38
UniGene ID:
Hs.652825
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1075
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALPK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102406.1
NP_001095876.1
16
4043
NM_001253884.1
NP_001240813.1
15
3888
NM_025144.3
NP_079420.3
16
4095
XM_005263245.4
XP_005263302.1
15
4192
XM_005263246.4
XP_005263303.1
15
4115
XM_006714326.3
XP_006714389.1
14
4132
XM_017008633.1
XP_016864122.1
15
4210
XM_017008634.1
XP_016864123.1
14
4178
XM_017008635.1
XP_016864124.1
15
4133
XM_017008636.1
XP_016864125.1
16
4286
XM_017008637.1
XP_016864126.1
15
4161
XM_017008638.1
XP_016864127.1
15
4299
XM_017008639.1
XP_016864128.1
16
4234
XM_017008640.1
XP_016864129.1
15
4109
XM_017008641.1
XP_016864130.1
15
4247
XM_017008642.1
XP_016864131.1
16
4113
XM_017008643.1
XP_016864132.1
14
4150
XM_017008644.1
XP_016864133.1
12
7343
XM_017008645.1
XP_016864134.1
15
4035
XM_017008646.1
XP_016864135.1
14
4087
XM_017008647.1
XP_016864136.1
14
4061
XM_017008648.1
XP_016864137.1
14
4009
XM_017008649.1
XP_016864138.1
10
3385
XM_017008650.1
XP_016864139.1
15
5795
XM_017008651.1
XP_016864140.1
15
4105
GenBank mRNA
AB040960.1
-
6
2488
AB075877.1
-
5
766
AK026715.1
-
5
776
AK097246.1
-
15
1922
AK131090.1
-
12
4305
AK304708.1
-
15
3851
AY044164.1
-
14
3898
BX648798.1
-
16
3986
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
protein phosphorylation
mitophagy in response to mitochondrial depolarization
xenophagy
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline