Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301073.1 | NP_001288002.1 | 4 | 660 | ||
NM_001301075.1 | NP_001288004.1 | 3 | 514 | |||
NM_001301077.1 | NP_001288006.1 | 4 | 572 | |||
NM_001301080.1 | NP_001288009.1 | 3 | 471 | |||
NM_001301082.1 | NP_001288011.1 | 3 | 184 | |||
NM_001301083.1 | NP_001288012.1 | 2 | 304 | |||
NM_004779.5 | NP_004770.4 | 4 | 708 | |||
XM_011537706.1 | XP_011536008.1 | 5 | 682 | |||
XM_011537708.2 | XP_011536010.1 | 3 | 185 | |||
XM_017010048.1 | XP_016865537.1 | 5 | 567 | |||
XM_017010049.1 | XP_016865538.1 | 5 | 1162 | |||
XM_017010050.1 | XP_016865539.1 | 5 | 1147 | |||
XM_017010051.1 | XP_016865540.1 | 5 | 1197 | |||
XM_017010057.1 | XP_016865546.1 | 5 | 725 | |||
GenBank mRNA | AF053318.1 | - | 4 | 582 | ||
AF087844.1 | - | 3 | 410 | |||
AF180476.1 | - | 3 | 410 | |||
AK025358.1 | - | 4 | 629 | |||
AK297381.1 | - | 3 | 379 | |||
AK299589.1 | - | 5 | 540 | |||
AK309359.1 | - | 4 | 462 | |||
AK314317.1 | - | 4 | 660 | |||
AK315864.1 | - | 3 | 363 | |||
AK316221.1 | - | 3 | 184 | |||
AK316269.1 | - | 2 | 302 | |||
AL122045.1 | - | 4 | 589 | |||
AL520837.3 | - | 5 | 803 | |||
BC017366.2 | - | 4 | 519 | 3506054 | ||
BT006857.1 | - | 3 | 338 | |||
BX647124.1 | - | 2 | 51 | |||
DC387160.1 | - | 4 | 440 |