Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004805.3 | NP_004796.1 | 4 | 1774 | ||
GenBank mRNA | BC002958.1 | - | 1 | 835 | ||
BC010427.1 | - | 1 | 835 | |||
BC017205.2 | - | 4 | 1745 | 3617979 | ||
U85510.1 | - | 4 | 1743 |