Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143970.1 | NP_001137442.1 | 7 | 2088 | ||
NM_001143971.1 | NP_001137443.1 | 7 | 2012 | |||
NM_004824.3 | NP_004815.3 | 7 | 2239 | |||
NR_026590.1 | 9 | 2194 | ||||
XM_011514997.2 | XP_011513299.1 | 7 | 2168 | |||
GenBank mRNA | AF081259.1 | - | 9 | 2194 | ||
AK291601.1 | - | 8 | 2002 | |||
AK296985.1 | - | 7 | 2088 | |||
AL050164.1 | - | 3 | 700 | |||
BC119682.2 | - | 6 | 1235 | 40116323 |