Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004996.3 | NP_004987.2 | 7 | 972 | ||
XM_011522497.1 | XP_011520799.1 | 7 | 788 | |||
XM_011522498.2 | XP_011520800.1 | 7 | 854 | |||
XM_017023237.1 | XP_016878726.1 | 7 | 854 | |||
XM_017023238.1 | XP_016878727.1 | 6 | 728 | |||
XM_017023239.1 | XP_016878728.1 | 6 | 716 | |||
XM_017023240.1 | XP_016878729.1 | 7 | 854 | |||
XM_017023241.1 | XP_016878730.1 | 5 | 590 | |||
XM_017023242.1 | XP_016878731.1 | 7 | 854 | |||
XM_017023243.1 | XP_016878732.1 | 7 | 854 | |||
GenBank mRNA | AB209120.1 | - | 5 | 492 | ||
AK311015.1 | - | 4 | 397 | |||
CR749835.1 | - | 3 | 250 | |||
L05628.1 | - | 7 | 993 | |||
X78338.1 | - | 7 | 934 |