Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282419.2 | NP_001269348.1 | 3 | 292 | ||
NM_001282420.2 | NP_001269349.1 | 4 | 354 | |||
NM_001282421.2 | NP_001269350.1 | 4 | 345 | |||
NM_001282422.2 | NP_001269351.1 | 3 | 220 | |||
NM_001291304.1 | NP_001278233.1 | 6 | 531 | |||
NM_005000.4 | NP_004991.1 | 4 | 337 | |||
NR_104168.2 | 5 | 546 | ||||
NR_104169.2 | 5 | 497 | ||||
NR_111925.1 | 4 | 335 | ||||
NR_111926.1 | 7 | 495 | ||||
GenBank mRNA | AK315458.1 | - | 4 | 248 | ||
AV713284.1 | - | 3 | 188 | |||
BC000813.1 | - | 4 | 253 | 3453236 | ||
BC020821.1 | - | 4 | 237 | 4701912 | ||
BC070236.1 | - | 4 | 238 | 4290594 | ||
BC070237.1 | - | 4 | 237 | 4659819 | ||
BF670375.1 | - | 488 | ||||
BF671339.1 | - | 5 | 395 | |||
BF790429.1 | - | 7 | 464 | |||
BG400546.1 | - | 5 | 448 | |||
BG611486.1 | - | 3 | 121 | |||
BT006695.1 | - | 4 | 228 | |||
CR933664.1 | - | 4 | 354 | |||
U53468.1 | - | 4 | 251 | |||
U64028.1 | - | 4 | 246 |