Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005071.2 | NP_005062.1 | 9 | 1691 | ||
NR_073589.1 | 9 | 1644 | ||||
XM_006722842.1 | XP_006722905.1 | 10 | 1874 | |||
XM_006722844.1 | XP_006722907.1 | 10 | 1767 | |||
XM_017027152.1 | XP_016882641.1 | 10 | 1962 | |||
GenBank mRNA | AK312996.1 | - | 9 | 1723 | ||
U18244.1 | - | 9 | 1691 |