Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 24 | 3431 | ||
NM_001145962.1 | NP_001139434.1 | 23 | 3354 | |||
NM_001145963.1 | NP_001139435.1 | 24 | 3328 | |||
NM_001145964.1 | NP_001139436.1 | 24 | 3319 | |||
NM_005072.4 | NP_005063.1 | 24 | 3449 | |||
GenBank mRNA | AF047338.1 | - | 24 | 3308 | ||
AF054505.1 | - | 23 | 3456 | |||
AF054506.1 | - | 23 | 3463 | |||
AK026911.1 | - | 16 | 2087 | |||
AK293906.1 | - | 24 | 3340 | |||
AK293956.1 | - | 23 | 3244 | |||
AK299042.1 | - | 24 | 3235 | |||
AK302790.1 | - | 24 | 3328 | |||
AK316415.1 | - | 24 | 3309 | |||
BC021193.2 | - | 24 | 3377 | 3349710 | ||
CF272417.1 | - | 2 | 170 | |||
U55054.1 | - | 24 | 3363 |