Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005074.3 | NP_005065.2 | 7, 8 | 859 | ||
XM_011514818.2 | XP_011513120.2 | 7, 8 | 896 | |||
XM_011514819.2 | XP_011513121.2 | 6, 7 | 809 | |||
XM_011514821.2 | XP_011513123.1 | 5, 6 | 613 | |||
XM_017011199.1 | XP_016866688.1 | 7, 8 | 896 | |||
XM_017011200.1 | XP_016866689.1 | 7, 8 | 896 | |||
XM_017011201.1 | XP_016866690.1 | 7, 8 | 896 | |||
XM_017011202.1 | XP_016866691.1 | 6, 7 | 824 | |||
GenBank mRNA | AK290783.1 | - | 7, 8 | 841 | ||
AK308035.1 | - | 7, 8 | 841 | |||
BC101745.1 | - | 6, 7 | 796 | 8069251 | ||
BC101747.1 | - | 6, 7 | 796 | 8069253 | ||
D28532.1 | - | 6, 7 | 740 | |||
X71355.1 | - | 6, 7 | 755 |