Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005116.5 | NP_005107.4 | 17 | 2154 | ||
NM_203327.1 | NP_976072.1 | 17 | 2172 | |||
XM_011529414.2 | XP_011527716.1 | 17 | 2159 | |||
XM_011529415.2 | XP_011527717.1 | 16 | 1980 | |||
XM_011529416.2 | XP_011527718.1 | 16 | 2289 | |||
XM_011529417.2 | XP_011527719.1 | 16 | 2036 | |||
XM_017028171.1 | XP_016883660.1 | 17 | 2193 | |||
XM_017028172.1 | XP_016883661.1 | 17 | 2197 | |||
XM_017028173.1 | XP_016883662.1 | 17 | 2177 | |||
XM_017028174.1 | XP_016883663.1 | 17 | 2072 | |||
XM_017028175.1 | XP_016883664.1 | 13 | 1508 | |||
XM_017028176.1 | XP_016883665.1 | 14 | 1826 | |||
XM_017028177.1 | XP_016883666.1 | 14 | 1813 | |||
XM_017028178.1 | XP_016883667.1 | 16 | 2079 | |||
XM_017028179.1 | XP_016883668.1 | 16 | 2066 | |||
XM_017028180.1 | XP_016883669.1 | 12 | 1390 | |||
GenBank mRNA | AF058319.1 | - | 15 | 1779 | ||
AF092511.1 | - | 16 | 2075 | |||
AF164142.1 | - | 17 | 2172 | |||
AJ269478.1 | - | 15 | 1779 | |||
AJ292318.1 | - | 16 | 2010 | |||
AK296304.1 | - | 14 | 1820 | |||
AY380556.1 | - | 15 | 1779 | |||
D87075.2 | - | 17 | 2154 | |||
EF032501.1 | - | 15 | 1779 |