Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042494.1 | NP_001035959.1 | 19 | 2714 | ||
NM_001042495.1 | NP_001035960.1 | 19 | 2389 | |||
NM_001042496.1 | NP_001035961.1 | 19 | 2780 | |||
NM_001042497.1 | NP_001035962.1 | 17 | 2483 | |||
NM_005135.2 | NP_005126.1 | 18 | 2353 | |||
NM_133647.1 | NP_598408.1 | 18 | 2528 | |||
XM_006720793.3 | XP_006720856.1 | 17 | 3263 | |||
GenBank mRNA | AF105366.1 | - | 18 | 2528 | ||
AF108831.1 | - | 18 | 2262 | |||
AF116242.1 | - | 18 | 2364 | |||
AF477977.1 | - | 19 | 2721 | |||
AF531258.1 | - | 17 | 2483 | |||
AF531259.1 | - | 19 | 2721 | |||
AF531260.1 | - | 19 | 2359 | |||
AK128133.1 | - | 17 | 2036 | |||
AK292550.1 | - | 19 | 2467 | |||
AK315283.1 | - | 18 | 2325 | |||
AL117500.1 | - | 15 | 1956 | |||
BC051744.1 | - | 19 | 2717 | 4829214 | ||
BC070107.1 | - | 19 | 2384 | 4826538 | ||
BC126241.1 | - | 18 | 2261 | 8991957 | ||
BC126243.1 | - | 18 | 2261 | 8991959 | ||
BX648195.1 | - | 18 | 3470 | |||
DQ138323.1 | - | 19 | 2779 |