Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130020.1 | NP_001123492.1 | 21 | 3190 | ||
NM_001130021.1 | NP_001123493.1 | 22 | 3187 | |||
NM_005177.3 | NP_005168.2 | 21 | 3169 | |||
XM_005257459.1 | XP_005257516.1 | 22 | 3190 | |||
XM_005257461.1 | XP_005257518.1 | 21 | 3018 | |||
XM_005257463.2 | XP_005257520.1 | 20 | 3000 | |||
XM_011524908.1 | XP_011523210.1 | 20 | 3064 | |||
XM_017024769.1 | XP_016880258.1 | 20 | 3027 | |||
XM_017024770.1 | XP_016880259.1 | 21 | 2981 | |||
XM_017024771.1 | XP_016880260.1 | 20 | 2963 | |||
XM_017024772.1 | XP_016880261.1 | 21 | 3000 | |||
XM_017024773.1 | XP_016880262.1 | 21 | 3581 | |||
XM_017024774.1 | XP_016880263.1 | 21 | 3163 | |||
GenBank mRNA | AK098213.1 | - | 2 | 927 | ||
AK125927.1 | - | 1 | 3700 | |||
AL096733.1 | - | 1 | 150 | |||
AL137683.1 | - | 4 | 1183 | |||
BC032398.1 | - | 22 | 3119 | 5195776 | ||
BX648978.1 | - | 21 | 3152 | |||
CR627443.1 | - | 15 | 2095 |