Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012732.1 | NP_001012750.1 | 6 | 1887 | ||
NM_001921.2 | NP_001912.2 | 6 | 1864 | |||
XM_005262778.3 | XP_005262835.1 | 6 | 1912 | |||
XM_005262779.3 | XP_005262836.1 | 7 | 2003 | |||
XM_005262780.3 | XP_005262837.1 | 8 | 2011 | |||
XM_005262781.4 | XP_005262838.1 | 5 | 1669 | |||
XM_005262782.3 | XP_005262839.1 | 5 | 1666 | |||
XM_006714115.3 | XP_006714178.1 | 6 | 1810 | |||
XM_006714116.3 | XP_006714179.1 | 5 | 2107 | |||
XM_011531674.2 | XP_011529976.1 | 7 | 2497 | |||
XM_011531675.2 | XP_011529977.1 | 6 | 2228 | |||
XM_011531676.2 | XP_011529978.1 | 7 | 2236 | |||
XM_017007816.1 | XP_016863305.1 | 7 | 1979 | |||
XM_017007817.1 | XP_016863306.1 | 7 | 1946 | |||
XM_017007818.1 | XP_016863307.1 | 8 | 2011 | |||
XM_017007819.1 | XP_016863308.1 | 8 | 2505 | |||
XM_017007820.1 | XP_016863309.1 | 8 | 2368 | |||
XM_017007821.1 | XP_016863310.1 | 8 | 2501 | |||
XM_017007822.1 | XP_016863311.1 | 8 | 1846 | |||
XM_017007823.1 | XP_016863312.1 | 5 | 1957 | |||
XM_017007824.1 | XP_016863313.1 | 5 | 1584 | |||
GenBank mRNA | BC088357.1 | - | 6 | 1778 | 30389169 | |
L12136.1 | - | 5 | 1690 |