Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 3 | 493 | ||
GenBank mRNA | AK075455.1 | - | 4 | 607 | ||
AK075481.1 | - | 4 | 394 | |||
AK293249.1 | - | 2 | 166 | |||
BC014433.1 | - | 3 | 434 | 3542405 | ||
BC036000.1 | - | 3 | 447 | 4712175 | ||
BC071878.1 | - | 3 | 436 | 6380973 | ||
D16234.1 | - | 3 | 412 | |||
D83485.1 | - | 3 | 367 | |||
EU668338.1 | - | 3 | 447 | |||
FJ224330.1 | - | 3 | 436 | |||
U42068.1 | - | 3 | 423 | |||
Z49835.1 | - | 3 | 451 |
Gene Symbol: PDIA3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002305.1 | 1 | 608 | |||
GenBank mRNA | AK295985.1 | - | 1 | 493 | ||
BM555251.1 | - | 1 | 552 |