Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005415.4 | NP_005406.3 | 8 | 1785 | ||
XM_017004768.1 | XP_016860257.1 | 8 | 1534 | |||
XM_017004769.1 | XP_016860258.1 | 9 | 4012 | |||
XM_017004770.1 | XP_016860259.1 | 9 | 1815 | |||
XM_017004771.1 | XP_016860260.1 | 10 | 1851 | |||
GenBank mRNA | AK123420.1 | - | 3 | 1323 | ||
AK312738.1 | - | 8 | 1704 | |||
BC019944.1 | - | 8 | 1692 | 3918690 | ||
BC075818.1 | - | 2 | 294 | |||
BX647555.1 | - | 1 | 1038 | |||
L20859.1 | - | 8 | 1616 |