Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001925.2 | NP_001916.1 | 3 | 554 | ||
GenBank mRNA | DN998607.1 | - | 3 | 503 | ||
X65977.1 | - | 3 | 480 |