Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145338.1 | NP_001138810.1 | 2 | 2572 | ||
NM_005453.4 | NP_005444.4 | 2 | 2590 | |||
GenBank mRNA | AK095978.1 | - | 1 | 2456 | ||
BC018541.1 | - | 2 | 2572 | 4385720 |