Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005471.4 | NP_005462.1 | 7 | 1657 | ||
XM_005268348.1 | XP_005268405.1 | 7 | 1914 | |||
XM_006714747.2 | XP_006714810.1 | 8 | 1681 | |||
XM_011537537.1 | XP_011535839.1 | 8 | 1976 | |||
GenBank mRNA | AF029914.1 | - | 6 | 1592 | ||
AJ002231.1 | - | 6 | 2338 | |||
AK022779.1 | - | 6 | 1516 | |||
BC001717.1 | - | 1 | 346 | |||
BC022322.1 | - | 7 | 1618 | 4243999 | ||
D31766.1 | - | 8 | 1980 |