Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321174.1 | NP_001308103.1 | 12 | 1348 | ||
NM_001321175.1 | NP_001308104.1 | 15 | 1520 | |||
NM_001321176.1 | NP_001308105.1 | 15 | 1515 | |||
NM_005486.2 | NP_005477.2 | 14 | 1512 | |||
XM_017024002.1 | XP_016879491.1 | 15 | 1432 | |||
GenBank mRNA | AJ010071.1 | - | 14 | 1367 | ||
AK001893.1 | - | 14 | 1416 | |||
AK223125.1 | - | 14 | 1346 | |||
AK300823.1 | - | 12 | 1169 | |||
AK303912.1 | - | 13 | 1266 | |||
AK303913.1 | - | 14 | 1329 | |||
AK315039.1 | - | 14 | 1350 | |||
AK315907.1 | - | 12 | 1186 |