Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 950 | ||
NM_001928.3 | NP_001919.2 | 5 | 929 | |||
GenBank mRNA | AK300963.1 | - | 5 | 924 | ||
BC034529.1 | - | 4 | 845 | |||
BC051001.1 | - | 5 | 869 | |||
BI763909.1 | - | 305 | ||||
M84526.1 | - | 5 | 877 |