Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 6 | 1558 | ||
XM_011542647.2 | XP_011540949.1 | 6 | 1205 | |||
GenBank mRNA | AK057299.1 | - | 6 | 1558 | ||
AK223596.1 | - | 6 | 947 | |||
AK296274.1 | - | 6 | 837 | |||
AK303930.1 | - | 7 | 783 | |||
AL832703.1 | - | 6 | 929 | |||
BC039084.1 | - | 6 | 927 | 4508489 | ||
Y00978.1 | - | 6 | 803 |