Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001935.3 | NP_001926.2 | 2, 3 | 660 | ||
GenBank mRNA | AB451339.1 | - | 1, 2 | 99 | ||
AB451488.1 | - | 1, 2 | 99 | |||
AY429531.1 | - | 1, 2 | 70 | |||
AY429532.1 | - | 1, 2 | 90 | |||
AY429533.1 | - | 1, 2 | 26 | |||
BC013329.2 | - | 1, 2 | 94 | |||
BC065265.1 | - | 2, 3 | 660 | 6156880 | ||
M74777.1 | - | 2, 3 | 109 | |||
M80536.1 | - | 1, 2 | 99 | |||
X60708.1 | - | 2, 3 | 174 |