Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134673.3 | NP_001128145.1 | 11 | 2841 | ||
NM_001145511.1 | NP_001138983.1 | 11 | 2685 | |||
NM_001145512.1 | NP_001138984.1 | 12 | 2574 | |||
NM_005595.4 | NP_005586.1 | 10 | 2749 | |||
XM_011541512.2 | XP_011539814.1 | 12 | 2956 | |||
XM_011541514.2 | XP_011539816.1 | 12 | 2889 | |||
XM_011541515.2 | XP_011539817.1 | 11 | 2864 | |||
XM_017001362.1 | XP_016856851.1 | 10 | 2671 | |||
XM_017001363.1 | XP_016856852.1 | 11 | 2332 | |||
GenBank mRNA | AB037860.1 | - | 11 | 2841 | ||
AK024964.1 | - | 4 | 1311 | |||
BC022264.1 | - | 10 | 2437 | 4270934 |