Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001938.2 | NP_001929.1 | 2 | 1042 | ||
GenBank mRNA | AK057541.1 | - | 3 | 904 | ||
AK311288.1 | - | 2 | 200 | |||
AL833729.1 | - | 2 | 1025 | |||
BC002809.2 | - | 2 | 1019 | 3634908 | ||
BC035507.2 | - | 2 | 1042 | 4555131 | ||
BC068553.1 | - | 2 | 1041 | 4822070 | ||
BT006972.1 | - | 2 | 317 | |||
M97388.1 | - | 3 | 864 |