Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141979.1 | NP_001135451.1 | 12 | 2704 | ||
NM_001141980.1 | NP_001135452.1 | 12 | 2704 | |||
NM_005657.2 | NP_005648.1 | 12 | 2724 | |||
XM_011521985.1 | XP_011520287.1 | 12 | 2703 | |||
XM_011521986.2 | XP_011520288.1 | 12 | 2703 | |||
XM_017022533.1 | XP_016878022.1 | 12 | 2659 | |||
GenBank mRNA | AB210025.1 | - | 12 | 2675 | ||
AF078776.1 | - | 12 | 2749 | |||
AK292915.1 | - | 12 | 2638 | |||
AK295764.1 | - | 12 | 2704 | |||
AK307405.1 | - | 12 | 2704 | |||
BC112161.1 | - | 12 | 2684 | 8327629 | ||
BX537418.1 | - | 12 | 2703 |