Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171184.1 | NP_001164655.1 | 3 | 457 | ||
NM_001939.2 | NP_001930.2 | 5 | 740 | |||
XM_017029333.1 | XP_016884822.1 | 5 | 898 | |||
XM_017029334.1 | XP_016884823.1 | 5 | 898 | |||
GenBank mRNA | AK289825.1 | - | 5 | 628 | ||
AK295843.1 | - | 3 | 326 | |||
BC111695.1 | - | 5 | 577 | 40036070 | ||
U43519.1 | - | 5 | 689 |