Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007026.1 | NP_001007027.1 | 4 | 467 | ||
NM_001940.3 | NP_001931.2 | 4 | 460 | |||
GenBank mRNA | BC051795.2 | - | 4 | 478 | 4181592 | |
BE142829.1 | - | 4 | 487 | |||
D31840.1 | - | 4 | 468 | |||
D38529.1 | - | 3 | 398 | |||
U23851.1 | - | 3 | 303 |