Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005689.2 | NP_005680.1 | 19 | 2895 | ||
GenBank mRNA | AF070598.1 | - | 15 | 1587 | ||
AF308472.1 | - | 19 | 2884 | |||
AJ289233.2 | - | 19 | 2855 | |||
AK026067.1 | - | 16 | 1704 | |||
AK057026.1 | - | 19 | 2614 | |||
AK172812.1 | - | 19 | 2245 | |||
BC000559.2 | - | 19 | 2846 | 3162881 | ||
BC043423.1 | - | 12 | 2895 | |||
U66673.1 | - | 8 | 877 |