Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 6 | 671 | ||
XM_005269379.2 | XP_005269436.1 | 6 | 731 | |||
GenBank mRNA | AK296669.1 | - | 5 | 588 | ||
AK312757.1 | - | 6 | 666 | |||
AL832720.1 | - | 1 | 3669 | |||
AY358854.1 | - | 1 | 806 | |||
BC005020.1 | - | 6 | 657 | 3659599 | ||
BC013204.1 | - | 6 | 648 | |||
BC110299.1 | - | 5 | 572 | 5215228 | ||
BE791220.1 | - | 6 | 686 | |||
M80254.1 | - | 6 | 669 |