Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 6 | 795 | ||
XM_005269379.2 | XP_005269436.1 | 6 | 855 | |||
GenBank mRNA | AK296669.1 | - | 5 | 712 | ||
AL832720.1 | - | 1 | 3793 | |||
AY358854.1 | - | 1 | 930 | |||
BC005020.1 | - | 6 | 781 | 3659599 | ||
BC013204.1 | - | 6 | 772 | |||
BC110299.1 | - | 5 | 696 | 5215228 | ||
M80254.1 | - | 6 | 793 |