Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 4 | 853 | ||
GenBank mRNA | AF057299.1 | - | 4 | 773 | ||
AF057300.1 | - | 4 | 708 | |||
AK290597.1 | - | 4 | 795 | |||
AK307439.1 | - | 4 | 528 | |||
BC062603.1 | - | 4 | 770 | |||
BC073850.1 | - | 4 | 770 | |||
BC108282.1 | - | 4 | 790 | |||
BC136436.1 | - | 4 | 745 | 9020425 | ||
DQ655928.2 | - | 4 | 456 | |||
U63139.1 | - | 4 | 840 |