Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 25 | 5671 | ||
GenBank mRNA | U63139.1 | - | 25 | 5658 |