Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190977.2 | NP_001177906.1 | 3 | 578 | ||
NM_001190979.2 | NP_001177908.1 | 5 | 776 | |||
NM_001320080.1 | NP_001307009.1 | 5 | 804 | |||
NM_005748.5 | NP_005739.2 | 4 | 704 | |||
NR_135139.1 | 7 | 946 | ||||
NR_135140.1 | 7 | 978 | ||||
XM_006719184.3 | XP_006719247.1 | 4 | 623 | |||
XM_006719185.3 | XP_006719248.1 | 4 | 648 | |||
XM_011537728.2 | XP_011536030.1 | 6 | 803 | |||
XM_011537729.2 | XP_011536031.1 | 4 | 593 | |||
XM_017018669.1 | XP_016874158.1 | 6 | 762 | |||
GenBank mRNA | AK291355.1 | - | 4 | 584 | ||
AK294260.1 | - | 3 | 499 | |||
AK296191.1 | - | 5 | 684 | |||
BC037777.2 | - | 5 | 644 | 5276663 | ||
BX649019.1 | - | 1 | 1013 |