Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243877.1 | NP_001230806.1 | 3 | 1978 | ||
NM_001243885.1 | NP_001230814.1 | 2 | 1789 | |||
NM_005749.3 | NP_005740.1 | 2 | 1992 | |||
GenBank mRNA | BC031406.1 | - | 1 | 1556 | 5194914 |