Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005762.2 | NP_005753.1 | 12 | 1842 | ||
GenBank mRNA | AK131063.1 | - | 5 | 430 | ||
AK225760.1 | - | 12 | 1842 | |||
AK313478.1 | - | 12 | 1559 | |||
BC004978.1 | - | 12 | 1842 | 2906325 | ||
BC007390.2 | - | 1 | 159 | |||
BC052986.2 | - | 10 | 1593 | 6502540 | ||
U31657.1 | - | 1 | 98 | |||
U78773.1 | - | 12 | 1653 | |||
U95040.1 | - | 12 | 1809 | |||
X97548.1 | - | 12 | 1914 |