Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005762.2 | NP_005753.1 | 13 | 2200 | ||
GenBank mRNA | AK131063.1 | - | 6 | 788 | ||
AK225760.1 | - | 13 | 2200 | |||
AK313478.1 | - | 13 | 1917 | |||
AL360114.1 | - | 1 | 26 | |||
AL360188.1 | - | 1 | 107 | |||
BC004978.1 | - | 13 | 2200 | 2906325 | ||
BC007390.2 | - | 2 | 517 | |||
BC052986.2 | - | 11 | 1951 | 6502540 | ||
U31657.1 | - | 2 | 456 | |||
U78773.1 | - | 13 | 2011 | |||
U95040.1 | - | 13 | 2167 | |||
X97548.1 | - | 13 | 2272 |