Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 8 | 1624 | ||
NM_001164194.1 | NP_001157666.1 | 8 | 1591 | |||
NM_005817.4 | NP_005808.3 | 8 | 1627 | |||
GenBank mRNA | AF051314.1 | - | 4 | 939 | ||
AF051315.1 | - | 4 | 915 | |||
AF055574.1 | - | 8 | 1503 | |||
AF057140.1 | - | 8 | 1524 | |||
AK122671.1 | - | 8 | 1478 | |||
AK225045.1 | - | 8 | 1493 | |||
AK225400.1 | - | 8 | 1508 | |||
AK226088.1 | - | 8 | 1505 | |||
AK291104.1 | - | 8 | 1584 | |||
BC001590.2 | - | 8 | 1500 | 2987965 | ||
BC005818.2 | - | 8 | 1516 | 3833411 | ||
BC007566.1 | - | 8 | 1516 | 3028104 | ||
BC019278.1 | - | 8 | 1516 | 2905275 |