Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005894.2 | NP_005885.1 | 6 | 2022 | ||
GenBank mRNA | U82812.1 | - | 6 | 1938 |