Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 6 | 860 | ||
NM_203364.2 | NP_976240.1 | 6 | 860 | |||
XM_017017749.1 | XP_016873238.1 | 5 | 831 | |||
GenBank mRNA | AK127972.1 | - | 5 | 660 | ||
BC001731.1 | - | 6 | 860 | 3355481 | ||
Z48042.1 | - | 5 | 713 |