Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003652.3 | NP_001003652.1 | 11 | 2121 | ||
NM_001135937.2 | NP_001129409.1 | 10 | 2031 | |||
NM_005901.5 | NP_005892.1 | 11 | 2013 | |||
XM_005258259.3 | XP_005258316.1 | 12 | 2254 | |||
XM_006722451.3 | XP_006722514.1 | 12 | 2266 | |||
XM_011525984.2 | XP_011524286.1 | 11 | 1610 | |||
XM_011525985.2 | XP_011524287.1 | 13 | 2313 | |||
XM_017025745.1 | XP_016881234.1 | 13 | 2352 | |||
XM_017025746.1 | XP_016881235.1 | 11 | 2164 | |||
XM_017025747.1 | XP_016881236.1 | 12 | 2239 | |||
XM_017025748.1 | XP_016881237.1 | 13 | 2325 | |||
XM_017025750.1 | XP_016881239.1 | 12 | 2214 | |||
GenBank mRNA | BC014840.1 | - | 11 | 1969 | 4449540 | |
BC025699.1 | - | 11 | 1827 | 5221801 | ||
BI258844.1 | - | 1 | 214 | |||
U59911.1 | - | 11 | 1843 | |||
U68018.1 | - | 10 | 1779 |