Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 2090 | ||
NM_001145103.1 | NP_001138575.1 | 9 | 1901 | |||
NM_001145104.1 | NP_001138576.1 | 7 | 1534 | |||
NM_005902.3 | NP_005893.1 | 9 | 2325 | |||
XM_011521559.2 | XP_011519861.1 | 8 | 2352 | |||
XM_011521560.1 | XP_011519862.1 | 9 | 2264 | |||
GenBank mRNA | AK026690.1 | - | 2 | 1723 | ||
AK301348.1 | - | 7 | 1418 | |||
BC050743.1 | - | 9 | 2243 | 6094328 | ||
CR749287.1 | - | 1 | 1086 | |||
U68019.1 | - | 10 | 2056 |