Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170931.1 | NP_001164402.1 | 4 | 2529 | ||
NM_005938.3 | NP_005929.2 | 3 | 2694 | |||
XM_011530957.2 | XP_011529259.1 | 3 | 2111 | |||
GenBank mRNA | BC026735.1 | - | 2 | 1803 | ||
X93996.1 | - | 3 | 2484 |