Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005955.2 | NP_005946.2 | 2 | 386 | ||
XM_011541491.2 | XP_011539793.1 | 2 | 550 | |||
XM_011541493.2 | XP_011539795.1 | 2 | 390 | |||
XM_011541494.1 | XP_011539796.1 | 2 | 358 | |||
GenBank mRNA | AK225923.1 | - | 2 | 358 | ||
AK309584.1 | - | 2 | 358 | |||
AK314233.1 | - | 2 | 358 | |||
AL702908.1 | - | 2 | 387 | |||
BC014454.1 | - | 2 | 361 | 4901099 | ||
X78710.1 | - | 2 | 328 |