Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005955.2 | NP_005946.2 | 11 | 2039 | ||
XM_011541491.2 | XP_011539793.1 | 11 | 2203 | |||
XM_011541494.1 | XP_011539796.1 | 12 | 2062 | |||
GenBank mRNA | AK225923.1 | - | 11 | 2011 | ||
AK298751.1 | - | 5 | 678 | |||
AK314233.1 | - | 11 | 2011 | |||
BC014454.1 | - | 11 | 2014 | 4901099 | ||
X78710.1 | - | 11 | 1981 |