Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321312.1 | NP_001308241.1 | 8 | 1827 | ||
NM_001321313.1 | NP_001308242.1 | 8 | 1613 | |||
NM_001321314.1 | NP_001308243.1 | 7 | 1533 | |||
NM_001321315.1 | NP_001308244.1 | 6 | 1446 | |||
NM_005966.3 | NP_005957.2 | 10 | 2089 | |||
XM_005246582.1 | XP_005246639.1 | 9 | 1762 | |||
XM_005246583.1 | XP_005246640.1 | 9 | 1876 | |||
XM_005246586.1 | XP_005246643.1 | 8 | 1566 | |||
XM_005246587.1 | XP_005246644.1 | 7 | 1479 | |||
XM_011511219.2 | XP_011509521.1 | 9 | 1731 | |||
XM_017004170.1 | XP_016859659.1 | 9 | 1772 | |||
XM_017004171.1 | XP_016859660.1 | 8 | 1939 | |||
XM_017004172.1 | XP_016859661.1 | 8 | 1711 | |||
XM_017004173.1 | XP_016859662.1 | 9 | 1794 | |||
XM_017004174.1 | XP_016859663.1 | 8 | 1610 | |||
XM_017004175.1 | XP_016859664.1 | 7 | 1688 | |||
XM_017004176.1 | XP_016859665.1 | 8 | 1681 | |||
GenBank mRNA | AF045451.1 | - | 7 | 1536 | ||
AF045452.1 | - | 6 | 1449 | |||
AK292446.1 | - | 8 | 1627 | |||
AK296549.1 | - | 6 | 774 | |||
BC035724.1 | - | 10 | 1978 | 5721972 | ||
U47007.1 | - | 7 | 1533 |