Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF1 SIRNA Assays ›
Gene Symbol:
IKZF1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location:
Chr.7: 50303453 - 50405101 on Build GRCh38
UniGene ID:
Hs.435949
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107931
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220765.2
NP_001207694.1
7
3350
NM_001220767.2
NP_001207696.1
6
2931
NM_001220768.2
NP_001207697.1
5
2961
NM_001220770.2
NP_001207699.1
5
2805
NM_001220771.2
NP_001207700.1
4
2793
NM_001291837.1
NP_001278766.1
7
3323
NM_001291838.1
NP_001278767.1
7
3188
NM_001291839.1
NP_001278768.1
6
2867
NM_001291840.1
NP_001278769.1
3
2532
NM_001291841.1
NP_001278770.1
5
2793
NM_001291842.1
NP_001278771.1
5
2763
NM_001291843.1
NP_001278772.1
4
2667
NM_001291844.1
NP_001278773.1
4
2637
NM_006060.5
NP_006051.1
8
3476
XM_011515058.2
XP_011513360.1
9
3366
XM_011515059.2
XP_011513361.1
9
3338
XM_011515060.2
XP_011513362.1
9
6516
XM_011515061.2
XP_011513363.1
8
3304
XM_011515062.2
XP_011513364.1
9
3646
XM_011515063.2
XP_011513365.1
9
3563
XM_011515064.2
XP_011513366.1
9
7194
XM_011515065.2
XP_011513367.1
9
3331
XM_011515066.2
XP_011513368.1
9
3435
XM_011515067.2
XP_011513369.1
8
3242
XM_011515068.2
XP_011513370.1
8
3503
XM_011515069.2
XP_011513371.1
8
7131
XM_011515070.2
XP_011513372.1
7
3181
XM_011515071.2
XP_011513373.1
8
3105
XM_011515072.2
XP_011513374.1
7
3043
XM_011515073.2
XP_011513375.1
8
3385
XM_011515074.2
XP_011513376.1
7
3016
XM_011515075.2
XP_011513377.1
7
3325
XM_011515076.2
XP_011513378.1
7
3269
XM_011515077.2
XP_011513379.1
6
2920
XM_011515078.2
XP_011513380.1
6
2893
XM_017011667.1
XP_016867156.1
8
3302
XM_017011668.1
XP_016867157.1
7
3009
GenBank mRNA
AF432219.1
-
2
2471
AY377974.1
-
7
3251
BC018349.1
-
7
3274
4338834
BU665456.1
-
1
38
JX459579.1
-
6
2623
U40462.1
-
8
3379
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
mesoderm development
covalent chromatin modification
lymphocyte differentiation
erythrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline