Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105529.1 | NP_001098999.1 | 12 | 1350 | ||
NM_006095.2 | NP_006086.1 | 12 | 1350 | |||
XM_005248043.2 | XP_005248100.1 | 12 | 1345 | |||
XM_011513615.1 | XP_011511917.1 | 12 | 1345 | |||
XM_011513616.2 | XP_011511918.1 | 12 | 1379 | |||
XM_011513618.1 | XP_011511920.1 | 12 | 1345 | |||
XM_017007645.1 | XP_016863134.1 | 12 | 1379 | |||
XM_017007646.1 | XP_016863135.1 | 11 | 1252 | |||
XM_017007647.1 | XP_016863136.1 | 11 | 1250 | |||
GenBank mRNA | AB013452.1 | - | 12 | 1131 | ||
AB209687.1 | - | 12 | 1203 | |||
AF067820.1 | - | 12 | 1118 | |||
BC109317.1 | - | 12 | 1112 | 40036536 | ||
BC109318.1 | - | 12 | 1118 | 40036538 |