Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006101.2 | NP_006092.1 | 13, 14 | 1639 | ||
GenBank mRNA | AF017790.1 | - | 13, 14 | 1561 | ||
AK289396.1 | - | 13, 14 | 1593 | |||
AK313184.1 | - | 13, 14 | 1591 | |||
BC005239.1 | - | 13, 14 | 1609 | |||
BC035617.1 | - | 13, 14 | 1570 | 5164424 |